
Fluid Thioglycollate Medium

Products
Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Chemical Composition
Capacity (mL)
Compatibility
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Fluid Thioglycollate Medium, GranuCult® prime, EP, USP, JP, ISO, granular, suitable for microbiology