Skip to Content
Merck
Flufenacet certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Flufenacet

Flufenacet certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
N-(4-Fluorophenyl)-N-isopropyl-2-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yloxy]acetamide
Empirical Formula (Hill Notation):
C14H13F4N3O2S
CAS Number:
Molecular Weight:
363.33

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique