![Fludioxonil metabolite CGA 192155 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/247/320/c9eade5b-aeb7-4f9b-b6c5-31df8022af66/640/c9eade5b-aeb7-4f9b-b6c5-31df8022af66.png)
Fludioxonil metabolite CGA 192155
![Fludioxonil metabolite CGA 192155 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/247/320/c9eade5b-aeb7-4f9b-b6c5-31df8022af66/640/c9eade5b-aeb7-4f9b-b6c5-31df8022af66.png)
Synonym(s):
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid
Empirical Formula (Hill Notation):
C8H4F2O4
CAS Number:
Molecular Weight:
202.11
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing