
Fenthionsulfoxide

Empirical Formula (Hill Notation):
C10H15O4PS2
CAS Number:
Molecular Weight:
294.33
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing