![Fenthion oxon sulfone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/180/439/2d2d9ed4-ac30-47ed-bcb6-057919953520/640/2d2d9ed4-ac30-47ed-bcb6-057919953520.png)
Fenthion oxon sulfone
![Fenthion oxon sulfone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/180/439/2d2d9ed4-ac30-47ed-bcb6-057919953520/640/2d2d9ed4-ac30-47ed-bcb6-057919953520.png)
Synonym(s):
Dimethyl 3-methyl-4-(methylsulfonyl)phenyl phosphate, Fenoxon sulfone
Empirical Formula (Hill Notation):
C10H15O6PS
CAS Number:
Molecular Weight:
294.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2