
Famotidine Related Compound F

Synonym(s):
3-{[2-(Diaminomethyleneamino)thiazol-4-yl}methylthio]propanoic acid
Empirical Formula (Hill Notation):
C8H12N4O2S2
CAS Number:
Molecular Weight:
260.34
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing