![Exo-1-Naphthyl Kwon [2.2.1] Bicyclic Phosphine](/deepweb/assets/sigmaaldrich/product/structures/376/476/5be457d0-df63-4a3d-823c-9a8002a7a813/640/5be457d0-df63-4a3d-823c-9a8002a7a813.png)
Exo-1-Naphthyl Kwon [2.2.1] Bicyclic Phosphine
![Exo-1-Naphthyl Kwon [2.2.1] Bicyclic Phosphine](/deepweb/assets/sigmaaldrich/product/structures/376/476/5be457d0-df63-4a3d-823c-9a8002a7a813/640/5be457d0-df63-4a3d-823c-9a8002a7a813.png)
Synonym(s):
(1S,4S,5R)-5-(naphthalen-1-yl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C22H22NO2PS
Molecular Weight:
395.45
Products
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1