Skip to Content
Merck
Etoposide British Pharmacopoeia (BP) Reference Standard

Etoposide

Etoposide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213, Etoposide
Empirical Formula (Hill Notation):
C29H32O13
CAS Number:
Molecular Weight:
588.56
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
pK Value
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade