![Etofylline European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/989/432/30da5153-dffe-41a5-8157-3c339e52a8b1/640/30da5153-dffe-41a5-8157-3c339e52a8b1.png)
Etofylline
![Etofylline European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/989/432/30da5153-dffe-41a5-8157-3c339e52a8b1/640/30da5153-dffe-41a5-8157-3c339e52a8b1.png)
Synonym(s):
7-(β-Hydroxyethyl)theophylline, 1,3-Dimethyl-7-(2-hydroxyethyl)xanthine
Empirical Formula (Hill Notation):
C9H12N4O3
CAS Number:
Molecular Weight:
224.22
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing