Skip to Content
Merck
Etidronic acid monohydrate ≥95.0% (T)

Etidronic acid monohydrate

Etidronic acid monohydrate ≥95.0% (T)
Synonym(s):
(1-Hydroxyethylidene)bisphosphonic acid monohydrate, 1-Hydroxyethylidenediphosphonic acid
Empirical Formula (Hill Notation):
C2H8O7P2 · H2O
CAS Number:
Molecular Weight:
224.04
EC Number:

Products

Product Category
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing