Skip to Content
Merck
Ethyl isovalerate natural, ≥98%, FCC, FG

Ethyl isovalerate

Ethyl isovalerate natural, ≥98%, FCC, FG
Linear Formula:
(CH3)2CHCH2COOC2H5
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing