Skip to Content
Merck
Ethylene oxide solution 2.5-3.3 M in THF

Ethylene oxide solution

Ethylene oxide solution 2.5-3.3 M in THF
Synonym(s):
Ethylene oxide solution
Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reaction Type
Reference Material Type
Special Grade
Technique