Skip to Content
Merck
Estradiol valerate European Pharmacopoeia (EP) Reference Standard

Estradiol Valerate

Estradiol valerate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
β-Estradiol 17-valerate, 1,3,5(10)-Estratriene-3,17β-diol 17-pentanote
Empirical Formula (Hill Notation):
C23H32O3
CAS Number:
Molecular Weight:
356.50

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing