
Eplerenone

Synonym(s):
9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic acid, gamma-lactone, methyl ester, Cgp 30083, Eplerenone [USAN], Epoxymexrenone, HSDB 7522, Inspra, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, gamma-lactone, methyl ester, (7alpha,11alpha,17alpha)-, SC-66110, Eplerenone
Empirical Formula (Hill Notation):
C24H30O6
CAS Number:
Molecular Weight:
414.49
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade