(−)-Epigallocatechin 3-gallate
Synonym(s):
(−)-Epigallocatechin gallate, (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG
Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing