Enalaprilat
Synonym(s):
Enalaprilat dihydrate, (2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid, Vasotec
Empirical Formula (Hill Notation):
C18H24N2O5 ·2H2O
CAS Number:
Molecular Weight:
384.42
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing