Skip to Content
Merck
Eicosapentaenoic Acid Ethyl Ester Pharmaceutical Secondary Standard: Certified Reference Material

Eicosapentaenoic Acid Ethyl Ester

Eicosapentaenoic Acid Ethyl Ester Pharmaceutical Secondary Standard: Certified Reference Material
Synonym(s):
cis-5,8,11,14,17-Eicosapentaenoic acid ethyl ester
Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1