Skip to Content
Merck
Efavirenz Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Efavirenz Related Compound A

Efavirenz Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(S)-1-(2-Amino-5-chlorophenyl)-1-(trifluoromethyl)-3-cyclopropyl-2-propyn-1-ol, (S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol
Empirical Formula (Hill Notation):
C13H11ClF3NO
CAS Number:
Molecular Weight:
289.68

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1