![ECO BRIJ® L4 average Mn ~362](/deepweb/assets/sigmaaldrich/product/structures/710/586/b64d896b-40a0-46b4-bc02-0dc782b381fb/640/b64d896b-40a0-46b4-bc02-0dc782b381fb.png)
ECO BRIJ® L4
![ECO BRIJ® L4 average Mn ~362](/deepweb/assets/sigmaaldrich/product/structures/710/586/b64d896b-40a0-46b4-bc02-0dc782b381fb/640/b64d896b-40a0-46b4-bc02-0dc782b381fb.png)
Synonym(s):
Brij® L4, Polyethylene glycol dodecyl ether, Polyoxyethylene (4) lauryl ether
Linear Formula:
(C20H42O5)n
CAS Number:
Products
Product Category
Brand
Sustainability
Feature
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing