Skip to Content
Merck
(+)-Tubocurarine chloride pentahydrate ≥97.0% (TLC)

(+)-Tubocurarine chloride pentahydrate

(+)-Tubocurarine chloride pentahydrate ≥97.0% (TLC)
Empirical Formula (Hill Notation):
C37H42Cl2N2O6 · 5H2O
CAS Number:
Molecular Weight:
771.72

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing