
Drometrizole trisiloxane

Synonym(s):
2-(2H-Benzotriazol-2-yl)-4-methyl-6-{2-methyl-3-{1,3,3,3-tetramethyl-1-[(trimethylsilyl)oxy]disiloxanyl}propyl}phenol, Silatrizole
Empirical Formula (Hill Notation):
C24H39N3O3Si3
CAS Number:
Molecular Weight:
501.84
Products
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1