
D-Ornithine monohydrochloride

Synonym(s):
(R)-2,5-Diaminopentanoic acid monohydrochloride
Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Amino Acid
Showing 1-1 of 1