
Docosahexaenoic Acid Ethyl Ester

Synonym(s):
4,7,10,13,16,19-Docosahexaenoic acid ethyl ester
Empirical Formula (Hill Notation):
C24H36O2
CAS Number:
Molecular Weight:
356.54
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2