Skip to Content
Merck
DMT-locG(ib) Phosphoramidite configured for PerkinElmer

DMT-locG(ib) Phosphoramidite

DMT-locG(ib) Phosphoramidite configured for PerkinElmer
Synonym(s):
DMT-locG(ib) amidite
Empirical Formula (Hill Notation):
C45H54N7O9P
CAS Number:
Molecular Weight:
867.93

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3