Skip to Content
Merck
Di-tert-butyldichlorosilane 98%

Di-tert-butyldichlorosilane

Di-tert-butyldichlorosilane 98%
Synonym(s):
DTBSCl2
Linear Formula:
[(CH3)3C]2SiCl2
CAS Number:
Molecular Weight:
213.22
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing