Skip to Content
Merck
Discovery® C18 Supelguard Guard Cartridge 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea

Discovery® C18 (5 µm) HPLC Columns

Discovery® C18 Supelguard Guard Cartridge 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2