![di-Phosphorus pentasulfide for synthesis](/deepweb/assets/sigmaaldrich/product/structures/190/507/56fa72de-145f-4eb3-84ae-e15cdfa7a01b/640/56fa72de-145f-4eb3-84ae-e15cdfa7a01b.png)
di-Phosphorus pentasulfide
![di-Phosphorus pentasulfide for synthesis](/deepweb/assets/sigmaaldrich/product/structures/190/507/56fa72de-145f-4eb3-84ae-e15cdfa7a01b/640/56fa72de-145f-4eb3-84ae-e15cdfa7a01b.png)
Synonym(s):
Phosphorus pentasulfide, Phosphorus(V) sulfide
Empirical Formula (Hill Notation):
P2S5
CAS Number:
Molecular Weight:
222.27
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing