
Diphenoxylate HCl

Synonym(s):
Diphenoxylate hydrochloride
Empirical Formula (Hill Notation):
C30H32N2O2 · HCl
CAS Number:
Molecular Weight:
489.05
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing