Skip to Content
Merck
Dimethyl phthalate-3,4,5,6-d4 analytical standard

Dimethyl phthalate-3,4,5,6-d4

Dimethyl phthalate-3,4,5,6-d4 analytical standard
Linear Formula:
C6D4(CO2CH3)2
CAS Number:
Molecular Weight:
198.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing