![Dienestrol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/780/dc6a869f-f530-4538-b489-bde320a0f69a/640/dc6a869f-f530-4538-b489-bde320a0f69a.png)
Dienestrol
![Dienestrol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/780/dc6a869f-f530-4538-b489-bde320a0f69a/640/dc6a869f-f530-4538-b489-bde320a0f69a.png)
Synonym(s):
3,4-Bis(4-hydroxyphenyl)-2,4-hexadiene
Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing