![Didanosine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/326/610/c06a2fba-9780-41e1-8bd4-3d24c9279bb9/640/c06a2fba-9780-41e1-8bd4-3d24c9279bb9.png)
Didanosine for system suitability
![Didanosine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/326/610/c06a2fba-9780-41e1-8bd4-3d24c9279bb9/640/c06a2fba-9780-41e1-8bd4-3d24c9279bb9.png)
Synonym(s):
2′,3′-Dideoxyinosine, ddI, ddIno
Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing