Skip to Content
Merck
Didanosine European Pharmacopoeia (EP) Reference Standard

Didanosine

Didanosine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2′,3′-Dideoxyinosine, ddI, ddIno
Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing