Skip to Content
Merck
D-Glucose-6,6-d2 API for Clinical Studies, 98 atom % D, 98% (CP)

D-Glucose-6,6-d2

D-Glucose-6,6-d2 API for Clinical Studies, 98 atom % D, 98% (CP)
Synonym(s):
Dextrose-6,6-d2
Empirical Formula (Hill Notation):
C6D2H10O6
CAS Number:
Molecular Weight:
182.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade