Skip to Content
Merck
D-Gluconic acid solution 49-53 wt. % in H2O

D-Gluconic acid solution

D-Gluconic acid solution 49-53 wt. % in H2O
Synonym(s):
2,3,4,5,6-Pentahydroxycaproic acid
Empirical Formula (Hill Notation):
C6H12O7
CAS Number:
Molecular Weight:
196.16

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing