
Dexketoprofen Tromethamine

Synonym(s):
Dexketoprofen tromethamine salt, (S)-3-Benzoyl-α-methylbenzeneacetic acid compound with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), (S)-Ketoprofen tromethamol, Dexketoprofen trometamol
Empirical Formula (Hill Notation):
C16H14O3 · C4H11NO3
CAS Number:
Molecular Weight:
375.42
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing