Skip to Content
Merck
Deoxycorticosterone acetate

Deoxycorticosterone acetate

Deoxycorticosterone acetate
Synonym(s):
11-Deoxycorticosterone acetate, 21-Acetoxy-4-pregnene-3,20-dione, 21-Acetoxyprogesterone, 21-Hydroxy-4-pregnene-3,20-dione 21-acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnen-21-ol-3,20-dione 21-acetate, Cortexone acetate, DOC acetate
Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
EC Number:

Products

Product Category
Biological Source
Formula Weight
Gene Alias
Physical Form
Research Area
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing