Skip to Content
Merck
D-Dihydrocarvone, mixture of isomers ≥97%, FCC, FG

(+)-Dihydrocarvone, mixture of isomers

D-Dihydrocarvone, mixture of isomers ≥97%, FCC, FG
Synonym(s):
p-Menth-8-en-2-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing