Skip to Content
Merck
Daidzin Pharmaceutical Secondary Standard; Certified Reference Material

Daidzin

Daidzin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4′,7-Dihydroxyisoflavone 7-glucoside, Daidzein 7-glucoside, Daidzein-7-O-β-D-glucopyranoside, Daidzin
Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4