![Cyclizine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/702/433/cc2f9ab1-8536-411e-9229-745b17c07fd9/640/cc2f9ab1-8536-411e-9229-745b17c07fd9.png)
Cyclizine impurity A
![Cyclizine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/702/433/cc2f9ab1-8536-411e-9229-745b17c07fd9/640/cc2f9ab1-8536-411e-9229-745b17c07fd9.png)
Synonym(s):
1-Methylpiperazine
Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing