![Cyasterone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/288/076/fe629e1d-82d9-467c-ab25-93593228f4a2/640/fe629e1d-82d9-467c-ab25-93593228f4a2.png)
Cyasterone
![Cyasterone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/288/076/fe629e1d-82d9-467c-ab25-93593228f4a2/640/fe629e1d-82d9-467c-ab25-93593228f4a2.png)
Synonym(s):
(2β,3β,5β,22R,24S,25S,28R)-2,3,14,20,22,28-Hexahydroxy-6-oxo-stigmast-7-en-26-oic acid γ-lactone
Empirical Formula (Hill Notation):
C29H44O8
CAS Number:
Molecular Weight:
520.65
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing