![Cyanidin 3-sambubioside chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/429/067/24c27241-b86e-4ceb-81aa-5d2db14e10b0/640/24c27241-b86e-4ceb-81aa-5d2db14e10b0.png)
Cyanidin 3-sambubioside chloride
![Cyanidin 3-sambubioside chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/429/067/24c27241-b86e-4ceb-81aa-5d2db14e10b0/640/24c27241-b86e-4ceb-81aa-5d2db14e10b0.png)
Synonym(s):
Cyanidin 3-(2-O-xylosylglucoside) chloride, Cyanidin 3-O-(2-O-β-D-xylopyranosyl)-β-D-glucopyranoside chloride
Empirical Formula (Hill Notation):
C26H29ClO15
CAS Number:
Molecular Weight:
616.95
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing