Crystal violet (C.I. 42555)
Synonym(s):
Crystal violet (C.I. 42555)
Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98
Products
Brand
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing