Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper/Beryllium
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF15572684
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.015 mm, size 150 × 150 mm, temper as rolled
Expand
View Pricing
GF82526801
Copper/Beryllium,
foil, Cu98%/Be 2%, thickness 0.015 mm, diameter 25 mm, temper as rolled
Expand
View Pricing
GF46586133
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.015 mm, 4 mm diameter, temper as rolled
Expand
View Pricing
GF05952933
Copper/Beryllium,
rod, Cu 98%/Be 2%, 25 mm diameter, length 200 mm, temper hard
Expand
View Pricing
GF12384402
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.004 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF06269935
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.004 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF13061685
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.38 mm, size 178 × 178 mm, temper annealed
Expand
View Pricing
GF21663525
Copper/Beryllium,
rod, Cu 98%/Be 2%, 25 mm diameter, length 50 mm, temper hard
Expand
View Pricing
GF57602673
Copper/Beryllium,
rod, Cu 98%/Be 2%, 20 mm diameter, length 50 mm, temper hard
Expand
View Pricing
GF41724581
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.015 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF48050153
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.004 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF85570803
Copper/Beryllium,
rod, Cu98%/Be 2%, 25 mm diameter, length 100 mm, temper hard
Expand
View Pricing
GF94447479
Copper/Beryllium,
foil, Cu98%/Be 2%, thickness 0.015 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF80700364
Copper/Beryllium,
rod, Cu98%/Be 2%, length 100 mm, 20 mm diameter, temper hard
Expand
View Pricing
GF63742709
Copper/Beryllium,
rod, 20 mm diameter, Cu98%/Be 2%, length 200 mm, temper hard
Expand
View Pricing
GF96573842
Copper/Beryllium,
rod, 20 mm diameter, Cu98%/Be 2%, length 500 mm, temper hard
Expand
View Pricing
GF98378735
Copper/Beryllium,
insulated wire, Cu98%/Be 2%, conductor diameter 0.075 mm, length 20 m, insulation thickness 0.008 mm
Expand
View Pricing
GF15572639
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.38 mm, size 89 × 89 mm, temper annealed
Expand
View Pricing
GF05933796
Copper/Beryllium,
foil, Cu 98%/Be 2%, thickness 0.015 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF70444245
Copper/Beryllium,
insulated wire, Cu98%/Be 2%, conductor diameter 0.075 mm, length 200 m, insulation thickness 0.008 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2