Skip to Content
Merck
Clotrimazole for peak identification European Pharmacopoeia (EP) Reference Standard

Clotrimazole for peak identification

Clotrimazole for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Clotrimazole, 1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole
Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1