Clopidogrel Related Compound A
Synonym(s):
(S)-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid hydrochloride
Empirical Formula (Hill Notation):
C15H14ClNO2S · HCl
CAS Number:
Molecular Weight:
344.26
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing