Clarithromycin Impurity Q
Synonym(s):
Clarithromycin N-oxide, 6-O-Methylerythromycin N-oxide
Empirical Formula (Hill Notation):
C38H69NO14
CAS Number:
Molecular Weight:
763.95
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing