Skip to Content
Merck
cis-3-Hexenyl butyrate natural, ≥95%, FG

cis-3-Hexenyl butyrate

cis-3-Hexenyl butyrate natural, ≥95%, FG
Linear Formula:
CH3CH2CH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
170.25
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing