Skip to Content
Merck
cis-3,7-Dimethyl-2,6-octadien-1-ol ≥97%, FCC, FG

cis-3,7-Dimethyl-2,6-octadien-1-ol

cis-3,7-Dimethyl-2,6-octadien-1-ol ≥97%, FCC, FG
Synonym(s):
Nerol
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing