![Cirsilineol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/895/675/0040fa99-c769-4e89-885b-a6a047e3e922/640/0040fa99-c769-4e89-885b-a6a047e3e922.png)
Cirsilineol
![Cirsilineol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/895/675/0040fa99-c769-4e89-885b-a6a047e3e922/640/0040fa99-c769-4e89-885b-a6a047e3e922.png)
Synonym(s):
4′,5-Dihydroxy-3′,6,7-trimethoxyflavone, 6-Methoxyluteolin 3′,7-dimethyl ether, Anisomelin, Eupatrin, Fastigenin
Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing