Cimetidine for peak identification
Synonym(s):
Cimetidine, SKF-92334
Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing